missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POGLUT1 Monoclonal Antibody (737512)

POGLUT1 Monoclonal Antibody (737512)
Brand: Invitrogen MA524350
This item is not returnable.
View return policy
Description
Specifications
| POGLUT1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q8NBL1 | |
| POGLUT1 | |
| CHO-derived recombinant human Protein O-Glucosyltransferase 1/POGLUT Arg24-Leu388. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin) | |
| 737512 | |
| Unconjugated | |
| POGLUT1 | |
| 9630046K23Rik; C3orf9; CAP10-like 46 kDa protein; CAP10-like protein; CLP46; hCLP46; hRumi; KDELC family like 1; KDELCL1; KTEL (Lys-Tyr-Glu-Leu) containing 1; KTEL motif-containing protein 1; KTELC1; MDS010; MDSRP; Myelodysplastic syndromes relative protein; O-glucosyltransferase Rumi homolog; Poglut1; Protein O-glucosyltransferase 1; protein O-xylosyltransferase; Protein O-xylosyltransferase POGLUT1; RGD1306248; Rumi; UNQ490/PRO1006; Wing-shaped neural plate protein; Wsnp; x 010 protein | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 56983 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction