missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POFUT2 Polyclonal Antibody

POFUT2 Polyclonal Antibody
Brand: Invitrogen PA542785
This item is not returnable.
View return policy
Description
POFUT2 Polyclonal Antibody for Western Blot, IHC
Specifications
| POFUT2 | |
| Polyclonal | |
| Unconjugated | |
| POFUT2 | |
| 2310011G23Rik; AI256847; BC003494; C21orf80; FUT13; GDP-fucose protein O-fucosyltransferase 2; KIAA0958; O-FucT-2; peptide-O-fucosyltransferase 2; POFUT2; protein O-fucosyltransferase 2 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 23275 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9Y2G5 | |
| POFUT2 | |
| Synthetic peptide directed towards the C-terminal of human POFUT2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction