missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ POFUT2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110192
This item is not returnable.
View return policy
Description
Immunogen sequence: MVFARHLREV GDEFRSRHLN STDDADRIPF QEDWMKMKVK LGSALGGPYL GVHLRRKDFI WGHRQDVPSL EGAVRKIRSL MKTHRLDKVF VATDAVRKEY EELKKLLPEM VRFEPTWEEL ELYKDGGVAI I.
Fucose is typically found as a terminal modification of branched chain glycoconjugates, but it also exists in direct O-linkage to serine or threonine residues within cystine knot motifs in epidermal growth factor-like repeats or thrombospondin type-1 repeats. POFUT2 is an O-fucosyltransferase that use THBS type-1 repeats as substrates.
Specifications
| POFUT2 | |
| Polyclonal | |
| Unconjugated | |
| POFUT2 | |
| 2310011G23Rik; AI256847; BC003494; C21orf80; FUT13; GDP-fucose protein O-fucosyltransferase 2; KIAA0958; O-FucT-2; peptide-O-fucosyltransferase 2; POFUT2; protein O-fucosyltransferase 2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 23275, 309686, 80294 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.53 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q8VHI3, Q9Y2G5 | |
| POFUT2 | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction