Learn More
Invitrogen™ POFUT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597014
Description
Immunogen sequence: SWDPAGYLLY CPCMGRFGNQ ADHFLGSLAF AKLLNRTLAV PPWIEYQHHK PPFTNLHVSY QKYFKLEPLQ AYHRVISLED FMEKLAPTHW PPEKRVAYCF EVAAQRSPDK KTCPMKEGNP FGPFWDQFHV SFNKSELFTG ISFSASYREQ WSQRRENHSC VTLLFPR; Positive Samples: HepG2, HL-60, Mouse heart, Mouse liver; Cellular Location: Endoplasmic reticulum.
POFUT1, an endoplasmic reticulum-residing member of the glycosyltransferase O-Fuc family, adds O-fucose through an O-glycosidic linkage to conserved serine or threonines in epidermal growth factor-like repeats of several cell surface and secreted proteins. Unlike its homolog POFUT2, POFUT1 can also catalyze the transfer of fucose to thrombospondin type 1 repeats. Many of the substrates of POFUT1 are involved in ligand-induced receptor signaling. One such protein is Notch; mouse ES cells lacking POFUT have normal levels of Notch receptors at the cell surface, but these receptors do not bind Notch ligands or exhibit Notch signaling.
Specifications
| POFUT1 | |
| Polyclonal | |
| Unconjugated | |
| POFUT1 | |
| DDD2; FUT12; GDP-fucose protein O-fucosyltransferase 1; KIAA0180; mKIAA0180; o-fucosyltransferase protein; O-Fuc-T; OFUCT1; O-FucT-1; O-FucT-1a; O-FucT-1b; O-FUT; peptide-O-fucosyltransferase 1; Pofut1; pofut1 {ECO:0000312; protein O-fucosyltransferase 1; protein O-fucosyltransferase 1 isoform 1 precursor; protein O-fucosyltransferase 1a; protein O-fucosyltransferase 1b; protein-O-fucosyltransferase 1; RGD:1303001}; zgc:110472 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 140484, 23509, 311551 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.71 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6EV70, Q91ZW2, Q9H488 | |
| POFUT1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 28-194 of human POFUT1 (NP_7584361). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.