missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ POFUT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119370
This item is not returnable.
View return policy
Description
POFUT1 Polyclonal Antibody for Western Blot, IHC (P)
POFUT1, an endoplasmic reticulum-residing member of the glycosyltransferase O-Fuc family, adds O-fucose through an O-glycosidic linkage to conserved serine or threonines in epidermal growth factor-like repeats of several cell surface and secreted proteins. Unlike its homolog POFUT2, POFUT1 can also catalyze the transfer of fucose to thrombospondin type 1 repeats. Many of the substrates of POFUT1 are involved in ligand-induced receptor signaling. One such protein is Notch; mouse ES cells lacking POFUT have normal levels of Notch receptors at the cell surface, but these receptors do not bind Notch ligands or exhibit Notch signaling.
Specifications
| POFUT1 | |
| Polyclonal | |
| Unconjugated | |
| POFUT1 | |
| DDD2; FUT12; GDP-fucose protein O-fucosyltransferase 1; KIAA0180; mKIAA0180; o-fucosyltransferase protein; O-Fuc-T; OFUCT1; O-FucT-1; O-FucT-1a; O-FucT-1b; O-FUT; peptide-O-fucosyltransferase 1; Pofut1; pofut1 {ECO:0000312; protein O-fucosyltransferase 1; protein O-fucosyltransferase 1 isoform 1 precursor; protein O-fucosyltransferase 1a; protein O-fucosyltransferase 1b; protein-O-fucosyltransferase 1; RGD:1303001}; zgc:110472 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 23509 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q9H488 | |
| POFUT1 | |
| Recombinant protein O-Fucosyltransferase 1. The antigen corresponds to amino acid range 238-388 of the target protein. | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction