missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POF1B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11398-1-AP-20UL
This item is not returnable.
View return policy
Description
Plays a key role in the organization of epithelial monolayers by regulating the actin cytoskeleton. May be involved in ovary development.Specifications
| POF1B | |
| Polyclonal | |
| Unconjugated | |
| POF1B | |
| POF, POF1B, premature ovarian failure, 1B, Protein POF1B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 302328, 69693, 79983 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Western Blot, Immunofluorescence | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8K4L4, Q8WVV4 | |
| Pof1b | |
| POF1B Fusion Protein Ag1945 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction