missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PODXL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16383-1-AP-150UL
This item is not returnable.
View return policy
Description
Podolyxin is a CD34 family member expressed by podocytes, vascular endothelium, mesothelium, and a subset of hematopoietic progenitors. It is the major sialoprotein in the glycocalyx of glomerular podocytes. The physiologic role of podolyxin in leukemic blasts is unknown. However, it is thought of as a candidate prostate cancer tumor aggressiveness gene. Podolyxin appears to complement CD34 as a useful cell surface marker for hemangioblasts, the common precursors of hematopoietic and endothelial cells.Specifications
| PODXL2 | |
| Polyclonal | |
| Unconjugated | |
| PODXL2 | |
| Endoglycan, PODLX2, podocalyxin like 2, Podocalyxin like protein 2, PODXL2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 297433, 319655, 50512 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.47 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8CAE9, Q9NZ53 | |
| Podxl2 | |
| PODXL2 Fusion Protein Ag9374 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction