Learn More
PODXL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca:  Proteintech 16383-1-AP-20UL
 vedi altre versioni di questo prodotto
Codice prodotto. 16805994
  • £135.00 / 20µL
Questo articolo non è restituibile. Consulta la politica di reso
Description
Description
Podolyxin is a CD34 family member expressed by podocytes, vascular endothelium, mesothelium, and a subset of hematopoietic progenitors. It is the major sialoprotein in the glycocalyx of glomerular podocytes. The physiologic role of podolyxin in leukemic blasts is unknown. However, it is thought of as a candidate prostate cancer tumor aggressiveness gene. Podolyxin appears to complement CD34 as a useful cell surface marker for hemangioblasts, the common precursors of hematopoietic and endothelial cells.
Specifications
Specifications
PODXL2
Polyclonal
Unconjugated
PODXL2
Endoglycan, PODLX2, podocalyxin like 2, Podocalyxin like protein 2, PODXL2
Rabbit
Antigen Affinity Chromatography
RUO
297433, 319655, 50512
-20°C
Liquid
Western Blot, Immunoprecipitation
0.47 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8CAE9, Q9NZ53
Podxl2
PODXL2 Fusion Protein Ag9374
20 μL
Primary
Human, Mouse, Rat