missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PODXL Monoclonal Antibody (5D4)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA515845

Product Code. 12887862

  • £431.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MA5-15845 targets PODXL in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15845 immunogen is purified recombinant fragment of human PODXL expressed in E. Coli. . MA5-15845 detects PODXL which has a predicted molecular weight of approximately 49kDa.

This gene encodes a member of the sialomucin protein family. The encoded protein was originally identified as an important component of glomerular podocytes. Podocytes are highly differentiated epithelial cells with interdigitating foot processes covering the outer aspect of the glomerular basement membrane. Other biological activities of the encoded protein include: binding in a membrane protein complex with Na+/H+ exchanger regulatory factor to intracellular cytoskeletal elements, playing a role in hematopoetic cell differentiation, and being expressed in vascular endothelium cells and binding to L-selectin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PODXL
Monoclonal
Conc. Not Determined
ascites with 0.03% sodium azide
O00592
PODXL
Purified recombinant fragment of human PODXL expressed in E. Coli.
RUO
5420
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ELISA, Western Blot
5D4
Unconjugated
PODXL
AW121214; GCTM-2 antigen; Gp200; Ly102; MGC138240; PC; PCLP; PCLP antibody; Pclp1; PCLP-1; PCLP1 antibody; PODLX2; podocalyxin; podocalyxin like; podocalyxin-like; podocalyxin-like protein 1; podolyxin; Podxl
Mouse
100 μL
Primary
Human
Antibody
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.