missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PODXL Monoclonal Antibody

Mouse Monoclonal Antibody
Brand: Invitrogen™ 433140
This item is not returnable.
View return policy
Description
PODXL Monoclonal Antibody for Western Blot, ICC/IF, IHC, Flow, IP
This gene encodes a member of the sialomucin protein family. The encoded protein was originally identified as an important component of glomerular podocytes. Podocytes are highly differentiated epithelial cells with interdigitating foot processes covering the outer aspect of the glomerular basement membrane. Other biological activities of the encoded protein include: binding in a membrane protein complex with Na+/H+ exchanger regulatory factor to intracellular cytoskeletal elements, playing a role in hematopoetic cell differentiation, and being expressed in vascular endothelium cells and binding to L-selectin.
Specifications
| PODXL | |
| Monoclonal | |
| Unconjugated | |
| PODXL | |
| AW121214; GCTM-2 antigen; Gp200; Ly102; MGC138240; PC; PCLP; PCLP antibody; Pclp1; PCLP-1; PCLP1 antibody; PODLX2; podocalyxin; podocalyxin like; podocalyxin-like; podocalyxin-like protein 1; podolyxin; Podxl | |
| Mouse | |
| IgM | |
| RUO | |
| 5420 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.4 | |
| O00592 | |
| PODXL | |
| Human embryonal carcinoma cell extract. | |
| 100 μg | |
| Primary | |
| Human, Baboon | |
| Antibody | |
| IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction