missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Podoplanin Mouse, Clone: PDPN/1433, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 10630-MSM1-P1
Additional Details : Weight : 0.01000kg
Specifications
Podoplanin | |
Cell Surface and Cytoplasmic | |
PDPN/1433 | |
Unconjugated | |
XP_006710358 | |
Mouse | |
38-43kDa | |
Protein A/G Purified | |
RUO | |
10630 | |
IgG1 κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
PDPN | |
Recombinant human Podoplanin (PDPN) protein fragment (aa24-126) | |
HeLa Cells. Angiosarcoma or Mesothelioma. | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |