missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Podoplanin Mouse anti-Human, BV421, Clone: LpMab-17, BD Horizon™
Mouse Monoclonal Antibody
Brand: BD Biosciences 566456
This item is not returnable.
View return policy
Description
Podoplanin Monoclonal antibody specifically detects Podoplanin in Human samples. It is validated for Flow Cytometry,Immunofluorescence,Immunohistochemistry (Frozen).
Specifications
| Podoplanin | |
| Monoclonal | |
| 0.2 mg/mL | |
| Mouse | |
| RUO | |
| Human | |
| Purified |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| LpMab-17 | |
| Brilliant Violet 421 | |
| 50 μg | |
| Primary | |
| Refrigerate (2 to 8°C) | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction