missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Podoplanin Antibody (PMab-1), mFluor Violet 610 SE, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP3-11972MFV610
This item is not returnable.
View return policy
Description
Podoplanin Monoclonal antibody specifically detects Podoplanin in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Neutralization
Specifications
| Podoplanin | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Neutralization | |
| PMab-1 | |
| Western Blot, ELISA, Immunohistochemistry, Block/Neutralize | |
| 36-KD, aggrus, Gp36, GP40, HT1A-1, hT1alpha-1, hT1alpha-2, lung type I cell membrane associated glycoprotein, lung type-I cell membrane-associated glycoprotein (T1A-2), OTS 8, OTS8, PA2.26 antigen, podoplanin, T1A, T1-alpha | |
| This antibody was raised by immunising rats with 14-residue synthetic peptide mpp3851, which corresponds to amino acids 38-51 (GDGMVPPGIEDKIT) of the platelet-aggregation-stimulating (PLAG) domain of mouse podoplanin. Spleen cells were then harvested, and fused to P3U1 cells to generate stable hybridomas. | |
| 0.1 mL | |
| Core ESC Like Genes, Signal Transduction, Stem Cell Markers | |
| 10630 | |
| Store at 4C in the dark. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction