missing translation for 'onlineSavingsMsg'
Learn More

Podoplanin Antibody (pmab-1), Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18426392 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18426392 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18426392 Supplier Novus Biologicals Supplier No. NBP203955AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Podoplanin Monoclonal antibody specifically detects Podoplanin in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen Podoplanin
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone pmab-1
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias 36-KD, aggrus, Gp36, GP40, HT1A-1, hT1alpha-1, hT1alpha-2, lung type I cell membrane associated glycoprotein, lung type-I cell membrane-associated glycoprotein (T1A-2), OTS 8, OTS8, PA2.26 antigen, podoplanin, T1A, T1-alpha
Host Species Rat
Immunogen Podoplanin
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10630
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.