missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Podocin/NPHS2 Peptide
Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Specifications
Specifications
| Gene ID (Entrez) | 7827 |
| Species | Human |
| Content And Storage | Store at -20°C. |
| For Use With (Application) | This peptide is useful as a blocking peptide for NBP2-26057. For further blocking peptide related protocol, click here . |
| Gene Alias | nephrosis 2, idiopathic, steroid-resistant (podocin), PDCNSRN1podocin |
| Gene Symbol | NPHS2 |
| Quantity | 100 μg |
| Research Category | Signal Transduction |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction