missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Podocalyxin Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18150-1-AP-20UL
This item is not returnable.
View return policy
Description
PODXL encodes a member of the sialomucin protein family. The encoded protein was originally identified as an important component of glomerular podocytes. Podocytes are highly differentiated epithelial cells with interdigitating foot processes covering the outer aspect of the glomerular basement membrane. Other biological activities of the encoded protein include: binding in a membrane protein complex with Na+/H+ exchanger regulatory factor to intracellular cytoskeletal elements, playing a role in hematopoetic cell differentiation, and being expressed in vascular endothelium cells and binding to L-selectin.Specifications
| Podocalyxin | |
| Polyclonal | |
| Unconjugated | |
| PODXL | |
| GCTM 2 antigen, Gp200, PC, PCLP, PCLP 1, PCLP1, Podocalyxin, podocalyxin like, Podocalyxin like protein 1, PODXL, RCC marker, Renal Cell Carcinoma marker, TRA 1 81,TRA 1 60, TRA-1-60 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 27205, 5420 | |
| -20°C | |
| Liquid |
| Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) | |
| 0.26 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O00592, Q9R0M4 | |
| Podxl | |
| Podocalyxin Fusion Protein Ag12844 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur