missing translation for 'onlineSavingsMsg'
Learn More

Podocalyxin Like Antibody (PcMab-47), DyLight 488, Novus Biologicals™

Product Code. 18363775 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18363775 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18363775 Supplier Novus Biologicals Supplier No. NBP314620G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Podocalyxin Like Monoclonal antibody specifically detects Podocalyxin Like in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Podocalyxin Like
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone PcMab-47
Conjugate DyLight 488
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias Gp200MGC138240, PCLP1, PCLP-1, PCLPGCTM-2 antigen, PCpodocalyxin, podocalyxin-like, Podocalyxin-like protein 1
Host Species Mouse
Immunogen Recombinant Podocalyxin Like ectodomain
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cellular Markers, Hematopoietic Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5420
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.