missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Podocalyxin Like Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 8 publications
£297.00 - £402.00
Specifications
| Antigen | Podocalyxin Like |
|---|---|
| Concentration | 0.3mg/mL |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18407090
|
Novus Biologicals
NBP1-83348-25ul |
25 μL |
£297.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18260927
|
Novus Biologicals
NBP1-83348 |
0.1 mL |
£402.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Podocalyxin Like Polyclonal specifically detects Podocalyxin Like in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Podocalyxin Like | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| Gp200MGC138240, PCLP1, PCLP-1, PCLPGCTM-2 antigen, PCpodocalyxin, podocalyxin-like, Podocalyxin-like protein 1 | |
| PODXL | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| 0.3mg/mL | |
| Polyclonal | |
| Rabbit | |
| Cancer, Cell Biology, Cellular Markers, Embryonic Stem Cell Markers, Hematopoietic Stem Cell Markers, Stem Cell Markers | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 5420 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:LPETMSSSPTAASTTHRYPKTPSPTVAHESNWAKCEDLETQTQSEKQLVLNLTGNTLCAGGASDEKLISLICRAVKATFNPAQDKCGIRLASVPGSQTVVVKEITIHTKLPAKDVYERLKDKWDELKEAGVSDMKLGD | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title