missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Podocalyxin Like Antibody (4F10), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-45168-0.1MG
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
Podocalyxin Like Monoclonal specifically detects Podocalyxin Like in Human, Rat, Rabbit samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications
| Podocalyxin Like | |
| Monoclonal | |
| 0.2mg/mL | |
| Flow Cytometry 5 - 10 ul/million cells in 0.1ml, Immunohistochemistry-Paraffin 1:50 - 1:100, Immunofluorescence 1:50 - 1:100 | |
| Gp200MGC138240, PCLP1, PCLP-1, PCLPGCTM-2 antigen, PCpodocalyxin, podocalyxin-like, Podocalyxin-like protein 1 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Podocalyxin is a member of the CD34 transmembrane sialomucin family. It is over-expressed on the podocyte foot projections and plays essential roles in kidney development and homeostasis, blood filtration and urine formation. It is also expressed on vascular endothelia, hematopoietic progenitors and a subset of neurons. Overexpression of podocalyxin may be linked to more aggressive tumor behavior. Podocalyxin antibody can identify podocytes in the urine (podocyturia) that may indicate glomerular disease, pre-eclampsia, and other kidney pathology. | |
| Store at 4C. | |
| IgM |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| 4F10 | |
| Unconjugated | |
| 10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
| PODXL | |
| A recombinant protein fragment containing the intracellular, transmembrane, and part of the extracellular domain of human podocalyxin. | |
| 0.1 mg | |
| Cancer, Cell Biology, Cellular Markers, Embryonic Stem Cell Markers, Hematopoietic Stem Cell Markers, Stem Cell Markers | |
| 5420 | |
| Human, Rat, Rabbit | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction