Learn More
Sampling Systems™ Pocket Sampler
Quick and easy to use sampler that has a GMP design, which means it can be stripped and cleaned very quickly. Sampling Systems™ Pocket Sampler is suitable for taking discrete, zone samples from free flowing powders and granules.
Marca: Sampling Systems™ 1220B-1500
Detalles adicionales : Weight : 5.50400kg
Descripción
- Made from high quality 316L stainless steel with PTFE or 316L Stainless Steel inner core
- For taking multi-level, discrete samples
- Suitable for free flowing powders
- Bayonet handle that allows for simple operation
- Sealing sleeve enables one pocket to be emptied at a time
- Samplers are supplied with pointed tips as standard
Especificaciones
Sampler | |
5 Pockets | |
20 mL | |
1500 mm |
Stainless Steel (316L) | |
Stainless Steel Inner Rod | |
25 mm |