missing translation for 'onlineSavingsMsg'
Learn More

Kern Sohn Pocket Balance CM 60-2N

Product Code. 17341563
Encompass_Preferred
Click to view available options
:
60 g
This item is not returnable. View return policy

Product Code. 17341563

Brand: Kern Sohn CM 602N

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Pocket scale with integrated calculator.

  • Simple and convenient 4-key operation
  • Hard case cover as protection against pressure and dust
  • Robust, handy, well-protected
  • Integrated calculator

Specifications

Capacity 60 g
Readability 0.01 g
Type Pocket Balance
Electrical Requirements Battery Operated
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.