missing translation for 'onlineSavingsMsg'
Learn More

Pocapavir, MedChemExpress

Product Code. 16496667
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16496667

Brand: Medchem Express HY104074

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pocapavir (SCH-48973) is an orally active capsid inhibitor. Pocapavir prevents virion uncoating upon entry into the cell. Pocapavir has antiviral activity against polioviruses. Pocapavir also inhibits enterovirus infections.

Pocapavir (SCH-48973) is an orally active capsid inhibitor. Pocapavir prevents virion uncoating upon entry into the cell. Pocapavir has antiviral activity against polioviruses. Pocapavir also inhibits enterovirus infections.

Specifications

Chemical Name or Material Pocapavir
CAS 146949-21-5
Color White
Purity Grade Notes Research
Molecular Formula C21H17Cl3O3
Quantity 5 mg
Synonym SCH-48973 V-073
Solubility Information DMSO : 100 mg/mL (236.00 mM; Need ultrasonic)
SMILES ClC1=C(OCC2=CC=C(COC3=CC=C(OC)C=C3Cl)C=C2)C(Cl)=CC=C1
Molecular Weight (g/mol) 423.72
Formula Weight 423.72
Percent Purity 98.55%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.