missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ POC1B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110191
This item is not returnable.
View return policy
Description
Immunogen sequence: TNFDELHCKG LTKRNLKRLH FDSPPHLLDI YPRTPHPHEE KVETVEINPK LEVIDLQIST PPVMDILSFD STTTTETSGR TLPDKGEEAC GYFLNPSLMS PECLPTTTKK KTEDMSDLPC ESQRSIPLAV TDALEHIMEQ LNVLTQTVSI LEQRLTLTED KLKDCLENQQ KLFSAVQQKS.
Required for ciliogenesis.
Specifications
| POC1B | |
| Polyclonal | |
| Unconjugated | |
| POC1B | |
| 4933430F16Rik; CORD20; Pix1; POC1 centriolar protein B; POC1 centriolar protein homolog B; POC1 centriolar protein homolog B (Chlamydomonas); POC1B; Proteome of centriole protein 1B; TUWD12; WD repeat domain 51B; WD repeat-containing protein 51B; WDR51B | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 282809 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.84 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8TC44 | |
| POC1B | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 299-478 of human POC1B (NP_758440.1). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction