missing translation for 'onlineSavingsMsg'
Learn More

PO43- f(P)-PHOSPHATE solution for Ion Chromatography 1g/L H2O matrix, Fisher chemical™
BIOPHARMA

Product Code. 12992175 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Packaging:
Bottle
Unit Size:
100mL
250mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
12992175 250 mL Bottle 250mL
12982175 100 mL Bottle 100mL
12902185 500 mL Bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12992175 Supplier Fisher Chemical Supplier No. A 037 250
  • ONLINE EXCLUSIVE PRICE
    £86.15
    £74.52 / 250mL
    Save £11.63
    13% Off
 Restricted Item
Estimated Shipment: 13-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Chemical Identifiers

CAS 7558-80-7
Molecular Formula H2NaO4P
Molecular Weight (g/mol) 119.98
MDL Number MFCD00003527,MFCD00146206
InChI Key AJPJDKMHJJGVTQ-UHFFFAOYSA-M
PubChem CID 23672064
ChEBI CHEBI:37585
SMILES [Na+].OP(O)([O-])=O

Specifications

Packaging Bottle
Physical Form Liquid
Concentration 1 g/L
Type Single-component Standard Solution
IUPAC Name sodium dihydrogen phosphate
Solvent or Matrix Water
Quantity 250 mL
Chemical Name or Material Phosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.