missing translation for 'onlineSavingsMsg'
Learn More

PNU-74654, MedChemExpress

Product Code. 16493278
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
25mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16493278 10 mg 10mg
16454536 100 mg 100mg
16440926 200 mg 200mg
16422706 25 mg 25mg
16416007 5 mg 5mg
16409826 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16493278 Supplier Medchem Express Supplier No. HY101130
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress PNU-74654 is an inhibitor of Wnt/β-catenin pathway with an IC50 of 129.8 μM in NCI-H295 cell.

PNU-74654 is an inhibitor of Wnt/β-catenin pathway with an IC50 of 129.8 μM in NCI-H295 cell.

Specifications

Chemical Name or Material PNU-74654
CAS 113906-27-7
Color White
Purity Grade Notes Research
Molecular Formula C19H16N2O3
Quantity 10 mg
Solubility Information DMSO : ≥ 30 mg/mL (93.65 mM)
SMILES O=C(N/N=C/C1=CC=C(C)O1)C2=CC=CC=C2OC3=CC=CC=C3
Molecular Weight (g/mol) 320.34
Formula Weight 320.34
Percent Purity 99.42%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.