missing translation for 'onlineSavingsMsg'
Learn More

PNU-74654, MedChemExpress

Product Code. 16416007
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16416007

Brand: Medchem Express HY101130

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PNU-74654 is an inhibitor of Wnt/β-catenin pathway with an IC50 of 129.8 μM in NCI-H295 cell.

PNU-74654 is an inhibitor of Wnt/β-catenin pathway with an IC50 of 129.8 μM in NCI-H295 cell.

Specifications

Chemical Name or Material PNU-74654
CAS 113906-27-7
Color White
Purity Grade Notes Research
Molecular Formula C19H16N2O3
Quantity 5 mg
Solubility Information DMSO : ≥ 30 mg/mL (93.65 mM)
SMILES O=C(N/N=C/C1=CC=C(C)O1)C2=CC=CC=C2OC3=CC=CC=C3
Molecular Weight (g/mol) 320.34
Formula Weight 320.34
Percent Purity 99.42%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.