missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PNU 74654

Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
Binds to β-catenin (KD = 450 nM). Inhibits the interaction between β-catenin and T cell factor 4 (Tcf4) and disrupts the Wnt signaling pathway.
Specifications
Specifications
| Chemical Name or Material | 2-Phenoxybenzoic acid-[(5-methyl-2-furanyl)methylene]hydrazide |
| CAS | 113906-27-7 |
| Assay Percent Range | >99% |
| Quantity | 25 mg |
| Molecular Formula | C19H16N2O3 |
| Formula Weight | Observed MW: 320.34 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction