missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PNU 74654

Description
Binds to β-catenin (KD = 450 nM). Inhibits the interaction between β-catenin and T cell factor 4 (Tcf4) and disrupts the Wnt signaling pathway.
Specifications
Specifications
| Chemical Name or Material | 2-Phenoxybenzoic acid-[(5-methyl-2-furanyl)methylene]hydrazide |
| CAS | 113906-27-7 |
| Assay Percent Range | >99% |
| Quantity | 25 mg |
| Molecular Formula | C19H16N2O3 |
| Formula Weight | Observed MW: 320.34 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?