missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ PNU 74654
GREENER_CHOICE

Product Code. 18703469
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 18703469

Brand: Tocris Bioscience™ 3534/5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

β-catenin binder; inhibits Wnt signaling

Binds to β-catenin (KD = 450 nM). Inhibits the interaction between β-catenin and T cell factor 4 (Tcf4) and disrupts the Wnt signaling pathway.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Phenoxybenzoic acid-[(5-methyl-2-furanyl)methylene]hydrazide
CAS 113906-27-7
Assay Percent Range >99%
Quantity 5 mg
Molecular Formula C19H16N2O3
Formula Weight Observed MW: 320.34
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.