missing translation for 'onlineSavingsMsg'
Learn More

PNU-282987 S enantiomer free base, MedChemExpress

Product Code. p-200096395
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
Unit Size:
2mg
5mg
This item is not returnable. View return policy

Product Code. 16497967

Brand: Medchem Express HY12560D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PNU-282987 S enantiomer free base is the S-enantiomer of PNU-282987 free base. PNU-282987 is an α7 nicotinic acetylcholine receptor (α7 nAChR) agonist.

PNU-282987 S enantiomer free base is the S-enantiomer of PNU-282987 free base. PNU-282987 is an α7 nicotinic acetylcholine receptor (α7 nAChR) agonist.

Specifications

Chemical Name or Material PNU-282987 S enantiomer free base
CAS 737727-12-7
Color White
Purity Grade Notes Research
Molecular Formula C14H17ClN2O
Quantity 5 mg
SMILES O=C(C1=CC=C(Cl)C=C1)N[C@H]2C3CCN(CC3)C2
Molecular Weight (g/mol) 264.75
Formula Weight 264.75
Percent Purity 99.58%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.