missing translation for 'onlineSavingsMsg'
Learn More

PNU-282987 S enantiomer free base, MedChemExpress

Product Code. 16497967
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
Unit Size:
2mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16497967 5 mg 5mg
16490166 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16497967 Supplier Medchem Express Supplier No. HY12560D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PNU-282987 S enantiomer free base is the S-enantiomer of PNU-282987 free base. PNU-282987 is an α7 nicotinic acetylcholine receptor (α7 nAChR) agonist.

PNU-282987 S enantiomer free base is the S-enantiomer of PNU-282987 free base. PNU-282987 is an α7 nicotinic acetylcholine receptor (α7 nAChR) agonist.

Specifications

Chemical Name or Material PNU-282987 S enantiomer free base
CAS 737727-12-7
Color White
Purity Grade Notes Research
Molecular Formula C14H17ClN2O
Quantity 5 mg
SMILES O=C(C1=CC=C(Cl)C=C1)N[C@H]2C3CCN(CC3)C2
Molecular Weight (g/mol) 264.75
Formula Weight 264.75
Percent Purity 99.58%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.