missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PNU 177864 hydrochloride, Tocris Bioscience™
Highly selective D3 antagonist
Chemical Identifiers
CAS | 250266-51-4 |
---|---|
Molecular Formula | C18H22ClF3N2O3S |
Molecular Weight (g/mol) | 438.89 |
InChI Key | UOMDLHQTLPWCCE-UHFFFAOYSA-N |
Synonym | pnu 177864 hydrochloride, n-4-2-propylamino ethyl phenyl-4-trifluoromethoxy-benzenesulfonamide hydrochloride |
PubChem CID | 56972219 |
IUPAC Name | N-[4-[2-(propylamino)ethyl]phenyl]-4-(trifluoromethoxy)benzenesulfonamide;hydrochloride |
SMILES | CCCNCCC1=CC=C(C=C1)NS(=O)(=O)C2=CC=C(C=C2)OC(F)(F)F.Cl |
Description
Highly selective dopamine D3 receptor antagonist. Induces phospholipidosis and exhibits antischizophrenic activity in vivo.Chemical Identifiers
250266-51-4 | |
438.89 | |
pnu 177864 hydrochloride, n-4-2-propylamino ethyl phenyl-4-trifluoromethoxy-benzenesulfonamide hydrochloride | |
N-[4-[2-(propylamino)ethyl]phenyl]-4-(trifluoromethoxy)benzenesulfonamide;hydrochloride |
C18H22ClF3N2O3S | |
UOMDLHQTLPWCCE-UHFFFAOYSA-N | |
56972219 | |
CCCNCCC1=CC=C(C=C1)NS(=O)(=O)C2=CC=C(C=C2)OC(F)(F)F.Cl |
Specifications
250266-51-4 | |
pnu 177864 hydrochloride, n-4-2-propylamino ethyl phenyl-4-trifluoromethoxy-benzenesulfonamide hydrochloride | |
CCCNCCC1=CC=C(C=C1)NS(=O)(=O)C2=CC=C(C=C2)OC(F)(F)F.Cl | |
438.89 | |
438.89 | |
PNU 177864 hydrochloride |
C18H22ClF3N2O3S | |
UOMDLHQTLPWCCE-UHFFFAOYSA-N | |
N-[4-[2-(propylamino)ethyl]phenyl]-4-(trifluoromethoxy)benzenesulfonamide;hydrochloride | |
56972219 | |
>99% |
Safety and Handling
Recommended Storage : Desiccate at Room Temperature