missing translation for 'onlineSavingsMsg'
Learn More

PNU-159682, MedChemExpress

Product Code. 16497138
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16497138

Brand: Medchem Express HY16700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PNU-159682, a metabolite of the anthracycline Nemorubicin, is a highly potent DNA topoisomerase II inhibitor with excellent cytotoxicity. PNU-159682 acts as a more potent and tolerated ADC cytotoxin than Doxorubicin for ADC synthesis. PNU-159682 can be used in EDV-nanocell technology to overcome drug resistance.

PNU-159682, a metabolite of the anthracycline Nemorubicin, is a highly potent DNA topoisomerase II inhibitor with excellent cytotoxicity. PNU-159682 acts as a more potent and tolerated ADC cytotoxin than Doxorubicin for ADC synthesis. PNU-159682 can be used in EDV-nanocell technology to overcome drug resistance.

Specifications

Chemical Name or Material PNU-159682
CAS 202350-68-3
Color Reddish Brown
Purity Grade Notes Research
Molecular Formula C32H35NO13
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (155.86 mM; Need ultrasonic)
SMILES COC1=C(C(C(C(O)=C([C@@H](O[C@H]2C[C@H]3[C@H](O[C@H]4N3CCO[C@@H]4OC)[C@H](C)O2)C[C@@](C(CO)=O)(O)C5)C5=C6O)=C6C7=O)=O)C7=CC=C1
Molecular Weight (g/mol) 641.62
Formula Weight 641.62
Percent Purity 95.11%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.