missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ PNU 120596
GREENER_CHOICE

Product Code. 18736696
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 18736696

Brand: Tocris Bioscience™ 2498/25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Positive allosteric modulator of α7 nAChRs; active in vivo

PNU 120596 is a positive allosteric modulator of α7 neuronal nicotinic acetylcholine receptors (EC50 = 216 nM), with no detectable effect on α4β2, α3β4 and α9α10 receptors. Active in vivo following systemic administration. Neuroprotective in an in vivo model of transient focal cerebral ischemia.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(5-Chloro-2,4-dimethoxyphenyl)-N'-(5-methyl-3-isoxazolyl)-urea
CAS 501925-31-1
Assay Percent Range >99%
Quantity 25 mg
Molecular Formula C13H14ClN3O4
Formula Weight Observed MW: 311.72
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.