missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PNR/NR2E3 Antibody (OTI3D4), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73510B
This item is not returnable.
View return policy
Description
PNR/NR2E3 Monoclonal specifically detects PNR/NR2E3 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| PNR/NR2E3 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| NR2E3 | |
| Full length recombinant protein of human PNR/NR2E3 (NP_057430) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot | |
| OTI3D4 | |
| Western Blot | |
| Nuclear receptor subfamily 2 group E member 3, nuclear receptor subfamily 2, group E, member 3, photoreceptor-specific nuclear receptor, PNRESCS, rd7, Retina-specific nuclear receptor, RNR, RP37MGC49976 | |
| Mouse | |
| Affinity Purified | |
| GPCR, Vision | |
| 10002.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction