missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PNPT1 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-22269FR
This item is not returnable.
View return policy
Description
PNPT1 Polyclonal antibody specifically detects PNPT1 in Human samples. It is validated for Western Blot
Specifications
| PNPT1 | |
| Polyclonal | |
| Western Blot | |
| DKFZp762K1914, EC 2.7.7, EC 2.7.7.8, mitochondrial, old-35, OLD35, PNPase, PNPase 1, PNPase old-35, Polynucleotide phosphorylase 1, Polynucleotide phosphorylase-like protein, polyribonucleotide nucleotidyltransferase 1 | |
| The immunogen this antibody was made to, maps to a region between residue 700 and 750 of human Polynucleotide Phosphorylase 1 using the numbering given in entry NP_149100.2 (GeneID 87178). | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 87178 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction