missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies pNPP ELISA Substrate

Product Code. 17110002
Encompass_Preferred
Click to view available options
Quantity:
120 mL
Unit Size:
120mL
This item is not returnable. View return policy

Product Code. 17110002

Brand: STEMCELL Technologies 01917

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Substrate for ELISA antibody pair kits

  • pNPP (para-nitrophenyl phosphate) is a chromogenic substrate for alkaline phosphatase (ALP) that is widely used in ELISA assays
  • pNPP ELISA Substrate is for use with STEMCELL's ELISA antibody pair kits and is supplied in a ready-to-use format in a diethanolamine buffer
  • The reaction of pNPP ELISA Substrate with ALP results in a yellow, water-soluble product with an absorbance that can be read at 405 nm

Specifications

Description B Cells; Hybridomas
Quantity 120 mL
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.