missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PNPO Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555687
This item is not returnable.
View return policy
Description
Immunogen sequence: MTCWLRGVTA TFGRPAEWPG YLSHLCGRSA AMDLGPMRKS YRGDREAFEE THLTSLDPVK QFAAWFEEAV QCPDIGEANA MCLATCTR Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 84%.
The enzyme encoded by this gene catalyzes the terminal, rate-limiting step in the synthesis of pyridoxal 5'-phosphate, also known as vitamin B6. Vitamin B6 is a required co-factor for enzymes involved in both homocysteine metabolism and synthesis of neurotransmitters such as catecholamine.
Specifications
| PNPO | |
| Polyclonal | |
| Unconjugated | |
| PNPO | |
| AI415282; epididymis secretory protein Li 302; FLJ10535; HEL-S-302; PDXPO; Pnpo; pyridoxal 5'-phosphate synthase; pyridoxamine 5'-phosphate oxidase; pyridoxamine-phosphate oxidase; pyridoxine 5'-phosphate oxidase; pyridoxine 5-phosphate oxidase; pyridoxine-5'-phosphate oxidase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 55163 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NVS9 | |
| PNPO | |
| Recombinant protein corresponding to Human PNPO. Recombinant protein control fragment (Product #RP-93192). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction