missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PNPO Antibody (OTI1G9), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73509H
This item is not returnable.
View return policy
Description
PNPO Monoclonal specifically detects PNPO in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| PNPO | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| PNPO | |
| Full length recombinant protein of human PNPO (NP_060599) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
| Western Blot, Immunohistochemistry | |
| OTI1G9 | |
| Western Blot, Immunohistochemistry | |
| EC 1.4.3.5, FLJ10535, PDXPO, pyridoxal 5′-phosphate synthase, pyridoxamine 5′-phosphate oxidase, Pyridoxamine-phosphate oxidase, pyridoxine 5′-phosphate oxidase, pyridoxine-5′-phosphate oxidase | |
| Mouse | |
| Affinity Purified | |
| Cancer, metabolism | |
| 55163.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction