missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PNPLA8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550992
This item is not returnable.
View return policy
Description
PNPLA8 Polyclonal Antibody detects endogenous levels of PNPLA8 protein.
Phospholipase A2 catalyzes cleavage of fatty acids from phospholipids, thereby regulating membrane physical properties and the release of lipid second messengers and growth factors. Phospholipase A2 activity also modulates cellular growth programs, inflammation, and ion channel function (Mancuso et al., 2000 [PubMed 10744668]).
Specifications
| PNPLA8 | |
| Polyclonal | |
| Unconjugated | |
| PNPLA8 | |
| 1200006O19Rik; AI467579; BM-043; Calcium-independent phospholipase A2-gamma; intracellular membrane-associated calcium-independent phospholipase A2 gamma; iPLA2 gamma; Ipla2(gamma); IPLA22; IPLA2-2; IPLA2G; iPLA2gamma; iPLA2-gamma; membrane-associated calcium-independent phospholipase A2 gamma; MMLA; patatin like phospholipase domain containing 8; patatin-like phospholipase domain containing 8; patatin-like phospholipase domain-containing protein 8; Pnpla8; PNPLA-gamma | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 50640, 67452 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8K1N1, Q9NP80 | |
| PNPLA8 | |
| Synthesized peptide derived from the C-terminal region of human iPLA2G. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction