missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PNPLA3 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA518901
This item is not returnable.
View return policy
Description
This antibody is predicted to react with canine, porcine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
The protein encoded by this gene is a triacylglycerol lipase that mediates triacylglycerol hydrolysis in adipocytes. The encoded protein, which appears to be membrane bound, may be involved in the balance of energy usage/storage in adipocytes.
Specifications
| PNPLA3 | |
| Polyclonal | |
| Unconjugated | |
| PNPLA3 | |
| 1-acylglycerol-3-phosphate O-acyltransferase Pnpla3; acylglycerol O-acyltransferase; Acylglycerol transacylase; Adiponutrin; ADPN; C22orf20; Calcium-independent phospholipase A2-epsilon; iPLA(2)epsilon; iPLA2epsilon; iPLA2-epsilon; Lysophosphatidic acid acyltransferase; patatin like phospholipase domain containing 3; patatin-like phospholipase domain containing 3; patatin-like phospholipase domain-containing protein 3; Pnpla3 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 116939, 80339 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q91WW7, Q9NST1 | |
| PNPLA3 | |
| Synthetic peptide sequence (EHDICPKVKSTN) corresponding to the internal amino acids of PNPLA3. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction