missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PNPLA3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117066
This item is not returnable.
View return policy
Description
Positive Samples: SKOV3, Mouse lung, Rat liver Immunogen sequence: PDMPDDVLWL QWVTSQVFTR VLMCLLPASR SQMPVSSQQA SPCTPEQDWP CWTPCSPKGC PAETKAEATP RSILRSSLNF FLGNKVPAGA EGLSTFPSFS LEKSL.
The protein encoded by this gene is a triacylglycerol lipase that mediates triacylglycerol hydrolysis in adipocytes. The encoded protein, which appears to be membrane bound, may be involved in the balance of energy usage/storage in adipocytes.
Specifications
| PNPLA3 | |
| Polyclonal | |
| Unconjugated | |
| PNPLA3 | |
| 1-acylglycerol-3-phosphate O-acyltransferase Pnpla3; acylglycerol O-acyltransferase; Acylglycerol transacylase; Adiponutrin; ADPN; C22orf20; Calcium-independent phospholipase A2-epsilon; iPLA(2)epsilon; iPLA2epsilon; iPLA2-epsilon; Lysophosphatidic acid acyltransferase; patatin like phospholipase domain containing 3; patatin-like phospholipase domain containing 3; patatin-like phospholipase domain-containing protein 3; Pnpla3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 116939, 362972, 80339 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.72 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q91WW7, Q9NST1 | |
| PNPLA3 | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction