missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress PNPG

Product Code. 16471579
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16471579 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16471579 Supplier MedChemExpress Supplier No. HY15927
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress PNPG(4-Nitrophenyl-β-D- glucopyranoside) is a chromogenic β-D-glucosidase substrate, producing a yellow solution upon cleavage.

Specifications

CAS 2492-87-7
Solubility DMSO : ≥ 48 mg/mL (159.34 mM) ∣H2O : 10 mg/mL (33.20 mM∣Need ultrasonic)
Synonym 4-Nitrophenyl β-D-glucopyranoside
Content And Storage -20°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
Quantity 1 g
Molecular Weight (g/mol) 301.25 g/mol
Product Type Biochemical Assay Reagents
Purity or Quality Grade 98.33%
Shipping Condition Room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.