missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PNPase Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA522396
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, Jurkat, Raji, mouse liver. Predicted reactivity: Human (99%), Mouse (93%), Rat (95%), Pig (94%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PNPT1 (polyribonucleotide nucleotidyltransferase 1, mitochondirla) is a RNA-binding protein implicated in numerous RNA metabolic processes. It catalyzes the phosphorolysis of single-stranded polyribonucleotides processively in the 3'-to-5' direction. PNPT1 is a component of the mitochondrial degradosome (mtEXO) complex that degrades 3' overhang double-stranded RNA with a 3'-to-5' directionality in an ATP-dependent manner. It is required for correct processing and polyadenylation of mitochondrial mRNAs. It plays a role as a cytoplasmic RNA import factor, in mitochondrial morphogenesis and respiration, regulation of expression of the electron transport chain, regulation of the stability of specific mature miRNAs in melanoma cells, and RNA cell surveilance. Mutations of the gene can result in combined oxidative phosphorylation deficiency 13 (COXPD13) and deafness, autosomal recessive, 70 (DFNB70).
Specifications
| PNPase | |
| Polyclonal | |
| Unconjugated | |
| PNPT1 | |
| 1200003F12Rik; 3'-5' RNA exonuclease OLD35; COXPD13; DFNB70; DKFZp762K1914; old 35; Old35; old-35; PNPase; PNPase 1; PNPase old-35; Pnpt1; Pnptl1; polynucleotide phosphorylase 1; Polynucleotide phosphorylase-like protein; polyribonucleotide nucleotidyltransferase 1; polyribonucleotide nucleotidyltransferase 1, mitochondrial | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 71701, 87178 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q8K1R3, Q8TCS8 | |
| PNPT1 | |
| Recombinant protein encompassing a sequence within the N-terminus region of human PNPase. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction