missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PNOC Monoclonal Antibody (CL15085)
Description
Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 76%.
PNOC is the ligand of the opioid receptor-like receptor (OPRL1). It may act as a transmitter in the brain by modulating nociceptive and locomotor behavior. This protein may be involved in neuronal differentiation and development.
Specifications
Specifications
| Antigen | PNOC |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | CL15085 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | PNOC |
| Gene Accession No. | Q13519, Q64387 |
| Gene Alias | N/OFQ; N23K; N23K/N27K; neuropeptide nociceptin 1; nociceptin; nociceptin/orphanin FQ precursor; nociceptin; prepronociceptin; nocistatin; NOP; Npnc1; OFQ; OFQ/N; ORL1 receptor agonist; Orphanin FQ; Orphanin FQ2; Pnoc; ppN/OFQ; PPNOC; pre-pro-N/OFQ; prepronociceptin; Prepronociceptin (neuropeptide nociceptin) (N23K); pronociceptin; proorphanin; propronociceptin |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?