missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PNMT Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562289
This item is not returnable.
View return policy
Description
Immunogen sequence: GHLLLIGALE ESWYLAGEAR LTVVPVSEEE VREALVRSGY KVRDLRTYIM PAHLQTGVDD VKGVFFAWAQ KV Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 92%.
The product of this gene catalyzes the last step of the catecholamine biosynthesis pathway, which methylates norepinephrine to form epinephrine (adrenaline). The enzyme also has beta-carboline 2N-methyltransferase activity. This gene is thought to play a key step in regulating epinephrine production.
Specifications
| PNMT | |
| Polyclonal | |
| Unconjugated | |
| Pnmt | |
| Noradrenaline N-methyltransferase; PENT; phenylethanolamine N-methylase; phenylethanolamine N-methyltransferase; phenylethanolamine-N-methyltransferase; PNMT; PNMTase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5409 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P11086 | |
| Pnmt | |
| Recombinant protein corresponding to Human PNMT. Recombinant protein control fragment (Product #RP-98466). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction