missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PNMT Monoclonal Antibody (OTI2D5)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525531
This item is not returnable.
View return policy
Description
PNMT Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
The product of this gene catalyzes the last step of the catecholamine biosynthesis pathway, which methylates norepinephrine to form epinephrine (adrenaline). The enzyme also has beta-carboline 2N-methyltransferase activity. This gene is thought to play a key step in regulating epinephrine production.
Specifications
| PNMT | |
| Monoclonal | |
| 0.7 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P11086 | |
| Pnmt | |
| Full length human recombinant protein of PNMT produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI2D5 | |
| Unconjugated | |
| Pnmt | |
| Noradrenaline N-methyltransferase; PENT; phenylethanolamine N-methylase; phenylethanolamine N-methyltransferase; phenylethanolamine-N-methyltransferase; PNMT; PNMTase | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5409 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction