missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PNMT Antibody (CL11434) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-21193JF525
This item is not returnable.
View return policy
Description
PNMT Monoclonal antibody specifically detects PNMT in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| PNMT | |
| Monoclonal | |
| Janelia Fluor 525 | |
| EC 2.1.1, EC 2.1.1.28, Noradrenaline N-methyltransferase, PENTMGC34570, phenylethanolamine N-methylase, phenylethanolamine N-methyltransferase, PNMTase | |
| This antibody was developed using a recombinant protein derived from P11086, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer | |
| 5409 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL11434 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction