missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PNMT Antibody (1C11), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-97942AF405
This item is not returnable.
View return policy
Description
PNMT Monoclonal specifically detects PNMT in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| PNMT | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PNMT | |
| Recombinant human PNMT (1-282aa) purified from E. coli | |
| 100 μL | |
| Cancer | |
| 5409 | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 1C11 | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| EC 2.1.1, EC 2.1.1.28, Noradrenaline N-methyltransferase, PENTMGC34570, phenylethanolamine N-methylase, phenylethanolamine N-methyltransferase, PNMTase | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Anti-human PNMT mAb, clone 1C11, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human PNMT protein. | |
| Store at 4C in the dark. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction