Learn More
Invitrogen™ PNMAL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110187
Description
Immunogen sequence: SREEARAQEA AEFEEMAAWA LAAGRKVKKE PGLAAEVGSA LKAETPNNWN ATEDQHEPTK PLVRRAGAKS RSRRKKQKKN SRQEAVPWKK PKGINSNSTA NLEDPEVGDA ESMAISEPIK GSRKPCVNKE ELALKKPMAK CAWKGPREPP QDARAEAESP GGASESDQDG GHESPPKKKA VAWVSAKNPA PMRKKKKVSL GPVSYVLVDS EDGRKKPVMP KKGPGSRREA SDQKAPRGQQ PAEATASTSR GPKAKPEGSP RRATNESRKV.
The function of this protein remains unknown.
Specifications
| PNMAL1 | |
| Polyclonal | |
| Unconjugated | |
| PNMA8A | |
| Paraneoplastic antigen-like protein 8A; paraneoplastic Ma antigen family like 1; paraneoplastic Ma antigen family-like 1; PNMA8A; PNMAL1; PNMA-like protein 1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 55228 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.7 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q86V59 | |
| PNMA8A | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 170-439 of human PNMAL1 (NP_060685.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.